Learn More
Aqua Solutions BROMOCRESOL GREEN .04% IN IPA
SDP
Catalog No. NC0473013
Encompass_Preferred
Bromocresol Green 0.04% w/v in IPA (1 liter flip top)
Catalog No. NC0473013 Supplier Aqua Solutions Supplier No. 13711LFT
Bromocresol Green 0.04% w/v in IPA (1 liter flip top)