Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Bromocresol green | 76-60-8 | 99.6% | 698.01 g/mol | C21H14Br4O5S | 1 ML
SDP

Catalog No. 5000209099
Encompass_Preferred

Bromocresol green is a pH-sensitive triphenylmethane dye used as a pH indicator and reagent for protein and albumin determination. It is supplied as a 10 mM solution in DMSO and packaged as a 1 mL vial.

  • pH-sensitive triphenylmethane dye.
  • Used for protein and albumin determination.
  • Formulated as 10 mM solution in DMSO, 1 mL.
  • Chemical formula C21H14Br4O5S; molecular weight 698.01 g/mol.
  • Purity 99.6%.
  • Recommended storage in solvent: -80°C (up to 2 years) or -20°C (up to 1 year).

Catalog No. 50-002-09099 Supplier Medchemexpress LLC Supplier No. HYW04014410MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Bromocresol green is a pH-sensitive triphenylmethane dye used as a pH indicator and reagent for protein and albumin determination. It is supplied as a 10 mM solution in DMSO and packaged as a 1 mL vial.

  • pH-sensitive triphenylmethane dye.
  • Used for protein and albumin determination.
  • Formulated as 10 mM solution in DMSO, 1 mL.
  • Chemical formula C21H14Br4O5S; molecular weight 698.01 g/mol.
  • Purity 99.6%.
  • Recommended storage in solvent: -80°C (up to 2 years) or -20°C (up to 1 year).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.