Learn More
Medchemexpress LLC Bromocresol green | 76-60-8 | 99.6% | 698.01 g·mol⁻¹ | C21H14Br4O5S | 100 G
SDP
Catalog No. 5000209646
Encompass_Preferred
Bromocresol green is a pH-sensitive triphenylmethane dye used as an analytical pH indicator and for determination of protein and albumin in serum. Supplied as a solid powder with reported high purity, it is provided with supporting documentation for laboratory use.
  • pH-sensitive triphenylmethane dye useful as a pH indicator and analytical reagent.
  • High reported purity suitable for analytical applications.
  • solid powder form for easy weighing and solution preparation.
  • available with datasheet, certificate of analysis, and safety data sheet.
  • registered under CAS number 76-60-8 for unambiguous identification.
Catalog No. 50-002-09646 Supplier Medchemexpress LLC Supplier No. HYW040144100G
Bromocresol green is a pH-sensitive triphenylmethane dye used as an analytical pH indicator and for determination of protein and albumin in serum. Supplied as a solid powder with reported high purity, it is provided with supporting documentation for laboratory use.
  • pH-sensitive triphenylmethane dye useful as a pH indicator and analytical reagent.
  • High reported purity suitable for analytical applications.
  • solid powder form for easy weighing and solution preparation.
  • available with datasheet, certificate of analysis, and safety data sheet.
  • registered under CAS number 76-60-8 for unambiguous identification.