Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Bromocresol green | 76-60-8 | MFCD00005874 | 95.0% | 698.01 g/mol | C21H14Br4O5S | 25 G
SDP

Catalog No. 5000209746
Encompass_Preferred

Bromocresol green is a pH-sensitive triphenylmethane dye used as a pH indicator and as a reagent for colorimetric determination of protein and albumin in serum. It is supplied in solid and solution forms for laboratory and analytical use.

  • pH indicator, changes from yellow to blue over pH 3.8-5.4.
  • Molecular weight 698.01 g/mol.
  • Chemical formula C21H14Br4O5S.
  • Available as solid packs (25 G, 50 G, 100 G) and as a 10 mM solution in DMSO.
  • Typical applications include titrations, culture media pH indication, and colorimetric protein assays.

Catalog No. 50-002-09746 Supplier Medchemexpress LLC Supplier No. 25GHYW040144
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Bromocresol green is a pH-sensitive triphenylmethane dye used as a pH indicator and as a reagent for colorimetric determination of protein and albumin in serum. It is supplied in solid and solution forms for laboratory and analytical use.

  • pH indicator, changes from yellow to blue over pH 3.8-5.4.
  • Molecular weight 698.01 g/mol.
  • Chemical formula C21H14Br4O5S.
  • Available as solid packs (25 G, 50 G, 100 G) and as a 10 mM solution in DMSO.
  • Typical applications include titrations, culture media pH indication, and colorimetric protein assays.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.