Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bromocresol Green Indicator, 0.04% w/v, Reagents

Catalog No. 01336569
Change view
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Glass Bottle
Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
01-336-569 100 mL Glass Bottle
01-336-568 500 mL Poly Bottle
01-336-567 500 mL Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01-336-569 Supplier Reagents Holdings Llc Supplier No. C7105500100C
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Bromocresol Green Indicator, 0.04% w/v, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Bromocresol Green Indicator, 0.04% w/v
CAS 7732-18-5, 62625-32-5
Boiling Point 100°C
Packaging Glass Bottle
Specific Gravity 1.001
Synonym Bromocresol blue, Bromcresol Green, Bromcresol Blue
Color Dark Blue
pH 3.8 to 5.4
Quantity 100 mL
Physical Form Liquid
Molecular Weight (g/mol) 705.96
Percent Purity 0.04%
Grade Reagent
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.