Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bromocresol Green Indicator, 1% (w/v), Ricca Chemical
SDP

Catalog No. R1209000500 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 L
Packaging:
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
R1209000500 500 mL Natural Poly Bottle
R12090001A 1 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. R1209000500 Supplier Ricca Chemical Company Supplier No. R1209000500A
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

Chemical Name or Material Bromocresol Green
CAS 7732-18-5, 62625-32-5
Odor Odorless
Packaging Natural Poly Bottle
Assay Percent Range 1% (w/v)
Color Dark Blue
pH >5
Solubility Information Solubility in Water: Infinite
Quantity 500 mL
Physical Form Solution
Percent Purity 1% w/v
Grade Indicator
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.