Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bromocresol green indicator, Reag. Ph. Eur., Solstice

Catalog No. 60046161 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-046-161 5 g Glass Bottle
60-046-160 25 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-046-161 Supplier Solstice Supplier No. 327425G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Reag. Ph. Eur.

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 76-60-8
Molecular Formula C21H14Br4O5S
Molecular Weight (g/mol) 698.014
MDL Number MFCD00005875
InChI Key FRPHFZCDPYBUAU-UHFFFAOYSA-N
Synonym 3′,3′′,5′,5′′-Tetrabromo-m-cresolsulfonephthalein, Bromocresol Blue
PubChem CID 6451
IUPAC Name 2,6-dibromo-4-[3-(3,5-dibromo-4-hydroxy-2-methylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-3-methylphenol
SMILES CC1=C(C(=C(C=C1C2(C3=CC=CC=C3S(=O)(=O)O2)C4=CC(=C(C(=C4C)Br)O)Br)Br)O)Br

Specifications

CAS Max % 100.0000%
Packaging Glass Bottle
Linear Formula C19H10Br4O5S
Beilstein 372527
Color Brown
Quantity 5 g
Formula Weight 698.01
Grade Ph. Eur.
Chemical Name or Material Bromocresol green indicator, Reag. Ph. Eur.
ShelfLife 1260 days from date of manufacture
Recommended Storage Room Temp
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.