Learn More
Sigma Aldrich Fine Chemicals Biosciences Bromocresol Green/Methl R500ML
Catalog No. 502900340
Encompass
Mixed indicator SOLUTIONons of bromocresol green (pk 4.9) and methyl red (pk 5) give a red color in acidic conditions and green color in basic conditions with a sharp transition through grey color at pH 5.1 as the two colors are complementary.
Catalog No. 50-290-0340 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 318728500ML
Mixed indicator SOLUTIONons of bromocresol green (pk 4.9) and methyl red (pk 5) give a red color in acidic conditions and green color in basic conditions with a sharp transition through grey color at pH 5.1 as the two colors are complementary.