Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bromocresol Green-Methyl Red Indicator in 50% Methanol, Reagents

Catalog No. 01335771
Change view
Click to view available options
Quantity:
120 mL
500 mL
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
01-335-771 500 mL Poly Bottle
01-335-772 120 mL Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-335-771 Supplier Reagents Holdings Llc Supplier No. CB113100500A
Add to Cart
Add to Cart

Bromocresol Green-Methyl Red Indicator in 50% Methanol, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Bromocresol Green-Methyl Red Indicator in 50% Methanol
CAS 7732-18-5, 67-56-1, 67-56-1
Synonym Bromcresol Green-Methyl Red, MRBG
Grade Reagent
Packaging Poly Bottle
Specific Gravity 0.9
Quantity 500 mL
Physical Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.