Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bromocresol Green, Methyl Red Indicator, APHA for Alkalinity, Alcoholic, Reagents

Catalog No. 01335847
Change view
Click to view available options
Quantity:
125 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-335-847 125 mL
01-335-845 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-335-847 Supplier Reagents Holdings Llc Supplier No. CB003500125A
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Bromocresol Green - Methyl Red Indicator APHA for Alkalinity, Alcoholic, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Bromocresol Green - Methyl Red Indicator APHA for Alkalinity, Alcoholic
Boiling Point 72°C
CAS 64-17-5, 67-63-0, 67-63-0
Packaging Poly Bottle
Specific Gravity 0.7907
Synonym Bromcresol Green-Methyl Red, MRBG
Quantity 125 mL
Physical Form Liquid
Grade Reagent
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.