Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bromocresol Green - Methyl Red Solution (Ethanol Soln.) [mixed Indicator], TCI America™
SDP

Catalog No. B0120500ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B0120500ML 500 mL
B0120100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B0120500ML Supplier TCI America Supplier No. B0120500ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

MDL Number MFCD00134126
Synonym Methyl Red - Bromocresol Green Solution

Specifications

CAS 64-17-5
UN Number 1170
Color Red
pH 5
Quantity 500 mL
Physical Form Liquid
Grade Indicator
Chemical Name or Material Bromocresol Green - Methyl Red Solution (Ethanol Soln.) [mixed Indicator]
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.