Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bromocresol Green, MP Biomedicals™

Catalog No. 0215250305 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
02-152-50305 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 02-152-50305 Supplier MP Biomedicals, Inc Supplier No. 0215250305
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Bromocresol Green is a tracking dye for DNA agarose electrophoresis and pH indicator.

  • Bromocresol Green is a dye of the triphenylmethane family (triarylmethane dyes).
  • It binds quantitatively with human albumin forming an intense blue-green complex with an absorbance maximum at 628 nm.
  • Bromocresol Green is useful in a variety of colorimetric detection technologies.
  • It is used as a tracking dye for DNA agarose gel electrophoresis, in protein determinations and in charge-transfer complexation processes.
  • In TLC, it is used for visualisation of the compounds with functional groups whose pKa is below 5.0.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 62625-32-5
Molecular Formula C21H13Br4NaO5S
Molecular Weight (g/mol) 720.00
InChI Key HEFSGAHJDGZCHA-UHFFFAOYNA-M
IUPAC Name sodium 2,6-dibromo-4-[3-(3,5-dibromo-4-hydroxy-2-methylphenyl)-1,1-dioxo-3H-2,1λ⁶-benzoxathiol-3-yl]-3-methylbenzen-1-olate
SMILES [Na+].CC1=C(Br)C(O)=C(Br)C=C1C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC(Br)=C([O-])C(Br)=C1C

Specifications

Melting Point 230°C (Decomposition)
pH 3.8 to 5.4
Quantity 5 g
Grade ACS
Chemical Name or Material Bromocresol Green
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.