Learn More
Abcam Bromocresol green, pH indicator, 5G
Catalog No. 502704719
Encompass_Preferred
MW 698.02 g/mol. Triiphenylmethane dye used as a pH indicator. Visual transition interval: pH 3.8 (yellow) – 5.4 (blue). Also used as a tracking dye for DNA agarose gel electrophoresis.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-4719 Supplier Abcam Supplier No. AB1463245G
MW 698.02 g/mol. Triiphenylmethane dye used as a pH indicator. Visual transition interval: pH 3.8 (yellow) – 5.4 (blue). Also used as a tracking dye for DNA agarose gel electrophoresis.
The product is subject to the following: Abcam Restricted Use Statement