Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bromocresol Green, Sigma-Aldrich

Catalog No. 01259082 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-259-082 5 g
01-259-086 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-259-082 Supplier MilliporeSigma Supelco Supplier No. 1143595G
Add to Cart
Add to Cart

Bromocresol green serves as a pH indicator in biology and medicinal experiments. It is used widely as a vital stain to study blood-brain permeability. It has been used in the pH-sensitive colorimetric determination of glutamate decarboxylase activity. It has also been employed in the evaluation of paper-analytical devices for colorimetric analysis of serum samples. It is used as a tracking dye for DNA agarose gel electrophoresis, in protein determinations, and in charge-transfer complexation processes. It is used in TLC for the visualization of compounds with functional groups whose pKa is below 5. 0.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Bromocresol Green
CAS 76-60-8
Molecular Weight (g/mol) 698.01 g/mol
Percent Purity 95%
Grade ACS
Color Blue to Green
pH 3.8 to 5.4
Quantity 5 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.