Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bromocresol Green Sodium Salt, Crystalline, Sigma-Aldrich

Catalog No. 01258853 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-258-853 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-258-853 Supplier MilliporeSigma Supelco Supplier No. B125625G
Add to Cart
Add to Cart

Crystalline pH indicator (≥98% purity) for diagnostic assay manufacturing, hematology, and histology.

Bromocresol Green sodium salt has been used as a pH indicator for glutamate decarboxylase activity assay in Listeria monocytogenes strains.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Bromocresol Green Sodium Salt
CAS 62625-32-5
Color Blue to Green
pH 3.8 to 5.4
Quantity 25 g
Molecular Weight (g/mol) 720 g/mol
Percent Purity ≥98%
Grade Indicator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.