Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bromocresol Green, Sodium Salt, MilliporeSigma™

Catalog No. 50947583 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-947-583 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-947-583 Supplier MilliporeSigma Supplier No. BX11603

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 62625-32-5
Molecular Weight (g/mol) 720

Specifications

Packaging Glass Bottle
Quantity 5 g
Recommended Storage Store at 15 to 30°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.