Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bromocresol Green, Sodium Salt, OmniPur™, MilliporeSigma™

Catalog No. 280025GM Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
28-002-5GM 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 28-002-5GM Supplier MilliporeSigma Supplier No. 280025GM

Chemical Identifiers

CAS 62625-32-5
Molecular Formula C21H13Br4NaO5S
Synonym 3′,3″,5′,5″-Tetrabromo-m-cresolsulfonphthalein sodium salt

Specifications

Packaging Bottle
Color Blue to Green
Quantity 25 g
Physical Form Powder or Crystals
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.