Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bromocresol Green Sodium Salt, Sigma-Aldrich

Catalog No. 01259008 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-259-008 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-259-008 Supplier MilliporeSigma Supelco Supplier No. 11436725G
Add to Cart
Add to Cart

90% (HPLC), ACS reagent, Dye content 90 %

Bromocresol Green (BCG), also known as bromcresol green, is a synthetic, anionic, triphenylmethane dye belonging to the phthalein and sulphonphthalein dyes group. It is commonly used as a pH indicator as it changes color from yellow to blue-green at pH 3. 8 to 5. 4. The monosodium salt of bromocresol green is highly ionizable in water. The dye at a higher pH level produces a large and weakly lipophilic dianion.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Bromocresol Green Sodium Salt
CAS 62625-32-5
Color Blue to Green
pH 3.8 to 5.4
Quantity 25 g
Molecular Weight (g/mol) 720 g/mol
Percent Purity 90%
Grade ACS
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.