Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Bromocresol Green Solution
SCI_ED

Catalog No. S25846 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S25846 100 mL
S25205 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S25846
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This product is for educational use - no C of A is available.

Specifications

Chemical Name or Material Bromcresol Green Solution
CAS 62625-32-5, 7732-18-5
Assay Percent Range 0.4% w/v
Linear Formula C24H43Br4NaO4S
Synonym BCG
Packaging 0.04% w/v Solution
Density 1.000
pH Color Changes pH 3.8-5.4 (yellow-blue)
Color Green
Quantity 100 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.