Learn More
Aqua Solutions BROMOCRESOL GRN-METHL
SDP
Catalog No. NC9143989
Encompass_Preferred
Bromocresol Green - Methyl Red Indic. Solution, Components: Water, Deionized ASTM Type II, CAS No.: 7732-18-5, Bromocresol Green Sodium Salt, CAS No.: 62625-32-5, 0.1%, Methyl Red Sodium Salt, CAS No.: 845-10-3, 0.02%, Melting Point: ~0degr
Catalog No. NC9143989 Supplier Aqua Solutions Supplier No. 1380100ML
Bromocresol Green - Methyl Red Indic. Solution, Components: Water, Deionized ASTM Type II, CAS No.: 7732-18-5, Bromocresol Green Sodium Salt, CAS No.: 62625-32-5, 0.1%, Methyl Red Sodium Salt, CAS No.: 845-10-3, 0.02%, Melting Point: ~0degr