Learn More
Medchemexpress LLC Bromocresol purple 100g
SDP
Catalog No. 5000206420
Encompass_Preferred
Bromocresol purple sodium is a pH indicator whose color changes from yellow to purple when the pH increases from 5 2 to 6 8 Bromocresol purple sodium can be used for detecting yeast cells with plasma membrane damage and measuring serum albumin concentrations[1][2][3]
Catalog No. 50-002-06420 Supplier Medchemexpress LLC Supplier No. HYW110790100G
Bromocresol purple sodium is a pH indicator whose color changes from yellow to purple when the pH increases from 5 2 to 6 8 Bromocresol purple sodium can be used for detecting yeast cells with plasma membrane damage and measuring serum albumin concentrations[1][2][3]