Learn More
Aat Bioquest Bromocresol purple [CAS 115-40-2] (Ultrapure grade)
SDP
Catalog No. 502979293
Encompass_Preferred
Bromocresol purple (BCP) or 5',5-dibromo-o-cresolsulfophthalein, is a pH indicator.
Catalog No. 50-297-9293 Supplier Aat Bioquest Supplier No. 92
Bromocresol purple (BCP) or 5',5-dibromo-o-cresolsulfophthalein, is a pH indicator.