Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Bromocresol purple | 115-40-2 | MFCD00011681 | >97.0% | 540.22 g/mol | C21H16Br2O5S | 50 MG
SDP

Catalog No. 5000208415
Encompass_Preferred

Bromocresol purple is a pH indicator dye supplied as a solid reagent for laboratory use. It changes color from yellow to purple across pH 5.2-6.8 and is commonly used for albumin assays and detecting yeast plasma membrane damage.

  • pH indicator range 5.2-6.8 (yellow to purple).
  • Used for serum albumin measurement.
  • Used to detect yeast plasma membrane damage.
  • Molecular weight 540.22 g/mol.
  • Chemical formula C21H16Br2O5S.
  • Pack size 50 mg.

Catalog No. 50-002-08415 Supplier Medchemexpress LLC Supplier No. HYW08792850MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Bromocresol purple is a pH indicator dye supplied as a solid reagent for laboratory use. It changes color from yellow to purple across pH 5.2-6.8 and is commonly used for albumin assays and detecting yeast plasma membrane damage.

  • pH indicator range 5.2-6.8 (yellow to purple).
  • Used for serum albumin measurement.
  • Used to detect yeast plasma membrane damage.
  • Molecular weight 540.22 g/mol.
  • Chemical formula C21H16Br2O5S.
  • Pack size 50 mg.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.