Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Bromocresol purple | 115-40-2 | MFCD00011681 | 99.4% | 540.22 g/mol | C21H16Br2O5S | 250 MG
SDP

Catalog No. 5000209306
Encompass_Preferred

Bromocresol purple is a pH indicator dye supplied as a solid reagent for laboratory use. It changes color from yellow to purple between pH 5.2 and 6.8 and is provided in a 250 mg pack suitable for buffer preparation and assay work.

  • pH transition range 5.2-6.8
  • High purity (99.4%)
  • Light brown solid appearance
  • Molecular formula C21H16Br2O5S
  • Molecular weight 540.22 g/mol
  • Available in a 250 mg pack size
  • Store at room temperature; in solvent store at -80°C for 2 years or -20°C for 1 year

Catalog No. 50-002-09306 Supplier Medchemexpress LLC Supplier No. 250MGHYW087928
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Bromocresol purple is a pH indicator dye supplied as a solid reagent for laboratory use. It changes color from yellow to purple between pH 5.2 and 6.8 and is provided in a 250 mg pack suitable for buffer preparation and assay work.

  • pH transition range 5.2-6.8
  • High purity (99.4%)
  • Light brown solid appearance
  • Molecular formula C21H16Br2O5S
  • Molecular weight 540.22 g/mol
  • Available in a 250 mg pack size
  • Store at room temperature; in solvent store at -80°C for 2 years or -20°C for 1 year

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.