Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bromocresol Purple Sodium Salt, Sigma-Aldrich

Catalog No. 01258950 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-258-950 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-258-950 Supplier MilliporeSigma Supelco Supplier No. 86089125G
Add to Cart
Add to Cart

Bromocresol Purple is widely used as a vital stain to trace fluid diffusion in the lymphatic system. In addition, the dye is suitable to differentiate dead or live yeast cells. As a pH indicator, Bromocresol Purple is used in microbial culture media, both broth, and agar. In analytical experiments, e. g. , to stain carbonic anhydrase isoenzyme on cellulose acetate electrophoresis membranes, bromocresol purple is used.

TRUSTED_SUSTAINABILITY

Specifications

CAS 62625-30-3
Color Brown to Red
Quantity 25 g
Molecular Weight (g/mol) 562.2 g/mol
Percent Purity 90%
Grade Indicator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.