Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bromocresol Purple Solution
SDPSCI_ED

Catalog No. S76762
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S76762 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S76762 Supplier Aqua Solutions Supplier No. 1390100ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Used as a narrow range pH indicator

Specifications

Chemical Name or Material Bromocresol Purple Solution
pH Color Changes pH 5.2-6.8 (yellow-purple)
Quantity 100 mL
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.