Learn More
Cayman Chemical Bromocriptine-13C-d3, Purity: ≥99% deuterated forms (d1-d3), 1 mg
SDP
Catalog No. 502934722
Encompass_Preferred
An internal standard for the quantification of bromocriptine by GC- or LC-MS
Catalog No. 50-293-4722 Supplier Cayman Chemical Supplier No. 266691 MG
An internal standard for the quantification of bromocriptine by GC- or LC-MS