Learn More
Apexbio Technology LLC Bromocriptine mesylate 22260-51-1 10mM (in 1mL DMSO)
SDP
Catalog No. 5000089725
Encompass_Preferred
Bromocriptine mesylate (CAS 22260-51-1) is a selective agonist of D -like dopamine receptors exhibiting high affinity with a pKi of 8 05 0 2 Through modulation of D receptor activity bromocriptine mesylate influences metabolic processes by reducing glucose intolerance and insulin resistance inhibits pituitary prolactin secretion and enhances motor function relevant to Parkinson s disease The compound has established applications in research on metabolic disorders neuroendocrine regulation and neurodegenerative diseases including studies pertaining to type 2 diabetes prolactinomas acromegaly and Parkinson s disease
Catalog No. 50-000-89725 Supplier Apexbio Technology LLC Supplier No. B62985.1
Bromocriptine mesylate (CAS 22260-51-1) is a selective agonist of D -like dopamine receptors exhibiting high affinity with a pKi of 8 05 0 2 Through modulation of D receptor activity bromocriptine mesylate influences metabolic processes by reducing glucose intolerance and insulin resistance inhibits pituitary prolactin secretion and enhances motor function relevant to Parkinson s disease The compound has established applications in research on metabolic disorders neuroendocrine regulation and neurodegenerative diseases including studies pertaining to type 2 diabetes prolactinomas acromegaly and Parkinson s disease