Learn More
Medchemexpress LLC Bromocriptine mesylate | 22260-51-1 | 99.9% | 10 MG
SDP
Catalog No. 5000342482
Encompass_Preferred
Bromocriptine mesylate is a potent dopamine D2/D3 receptor agonist that binds to the D2 dopamine receptor with a pKi of 8.05. It is permeable to the blood-brain barrier.
  • Potent dopamine D2/D3 receptor agonist
  • Binds D2 dopamine receptor with pKi of 8.05
  • Permeable to the blood-brain barrier
  • Stimulates [35S]-GTPγS binding at D2 dopamine receptor expressed in CHO cells with pEC50 of 8.15±0.05
  • Strong inhibitor of brain nitric oxide synthase (NOS) (IC50=10±2 μM)
  • Poorly active towards inducible macrophage NOS (IC50>100 μM)
  • Potent inhibitor of CYP3A4 with a calculated IC50 value of 1.69 μM
  • Demonstrates significant anti-immobility action in forced swimming test (FST) and tail suspension test (TST) in mice
  • Potentiates the anti-immobility action of MPE
  • Decreases static mechanical allodynia (SMA) score in rats
  • Induces a significant, dose-dependent decrease in pain scores in the CCI-IoN group
Catalog No. 50-003-42482 Supplier Medchemexpress LLC Supplier No. HY12705A10MG
Bromocriptine mesylate is a potent dopamine D2/D3 receptor agonist that binds to the D2 dopamine receptor with a pKi of 8.05. It is permeable to the blood-brain barrier.
  • Potent dopamine D2/D3 receptor agonist
  • Binds D2 dopamine receptor with pKi of 8.05
  • Permeable to the blood-brain barrier
  • Stimulates [35S]-GTPγS binding at D2 dopamine receptor expressed in CHO cells with pEC50 of 8.15±0.05
  • Strong inhibitor of brain nitric oxide synthase (NOS) (IC50=10±2 μM)
  • Poorly active towards inducible macrophage NOS (IC50>100 μM)
  • Potent inhibitor of CYP3A4 with a calculated IC50 value of 1.69 μM
  • Demonstrates significant anti-immobility action in forced swimming test (FST) and tail suspension test (TST) in mice
  • Potentiates the anti-immobility action of MPE
  • Decreases static mechanical allodynia (SMA) score in rats
  • Induces a significant, dose-dependent decrease in pain scores in the CCI-IoN group