Learn More
Medchemexpress LLC Bromocriptine mesylate | 22260-51-1 | 99.9% | 750.70 | 50 MG
SDP
Catalog No. 5000364531
Encompass_Preferred
Bromocriptine mesylate is a potent dopamine D2/D3 receptor agonist, binding to the D2 dopamine receptor with a pKi of 8.05. It is permeable to the blood-brain barrier. It stimulates [35S]-GTPγS binding at D2 dopamine receptors and acts as a strong inhibitor of brain nitric oxide synthase (neuronal NOS) and a potent inhibitor of CYP3A4.
  • Potent dopamine D2/D3 receptor agonist
  • Permeable to the blood-brain barrier
  • Stimulates [35S]-GTPγS binding at D2 dopamine receptors
  • Strong inhibitor of brain nitric oxide synthase (neuronal NOS)
  • Potent inhibitor of CYP3A4
Catalog No. 50-003-64531 Supplier Medchemexpress LLC Supplier No. 50MGHY12705A
Bromocriptine mesylate is a potent dopamine D2/D3 receptor agonist, binding to the D2 dopamine receptor with a pKi of 8.05. It is permeable to the blood-brain barrier. It stimulates [35S]-GTPγS binding at D2 dopamine receptors and acts as a strong inhibitor of brain nitric oxide synthase (neuronal NOS) and a potent inhibitor of CYP3A4.
  • Potent dopamine D2/D3 receptor agonist
  • Permeable to the blood-brain barrier
  • Stimulates [35S]-GTPγS binding at D2 dopamine receptors
  • Strong inhibitor of brain nitric oxide synthase (neuronal NOS)
  • Potent inhibitor of CYP3A4