Learn More
Medchemexpress LLC Bromocriptine mesylate | 22260-51-1 | 99.90% | 1 ML
SDP
Catalog No. 5000340035
Encompass_Preferred
Bromocriptine mesylate is a potent dopamine D2/D3 receptor agonist that readily crosses the blood-brain barrier. It stimulates [35S]-GTPγS binding at the D2 dopamine receptor expressed in CHO cells with a pEC50 of 8.15±0.05. It also acts as a strong inhibitor of brain nitric oxide synthase and a potent inhibitor of CYP3A4.
  • It is a potent dopamine D2/D3 receptor agonist.
  • It is permeable to the blood-brain barrier.
  • It shows significant anti-immobility action in experimental models of depression.
  • It induces a significant, dose-dependent decrease in pain scores in specific models.
Catalog No. 50-003-40035 Supplier Medchemexpress LLC Supplier No. HY12705A10MM/1ML
Bromocriptine mesylate is a potent dopamine D2/D3 receptor agonist that readily crosses the blood-brain barrier. It stimulates [35S]-GTPγS binding at the D2 dopamine receptor expressed in CHO cells with a pEC50 of 8.15±0.05. It also acts as a strong inhibitor of brain nitric oxide synthase and a potent inhibitor of CYP3A4.
  • It is a potent dopamine D2/D3 receptor agonist.
  • It is permeable to the blood-brain barrier.
  • It shows significant anti-immobility action in experimental models of depression.
  • It induces a significant, dose-dependent decrease in pain scores in specific models.