Learn More
Medchemexpress LLC Bromocriptine mesylate | 22260-51-1 | MFCD00069218 | 100.0% | 750.70 g/mol | C33H44BrN5O8S | 5 MG
SDP
Catalog No. 5000264443
Encompass_Preferred
Bromocriptine mesylate is the methanesulfonate salt of bromocriptine, a semisynthetic ergot-derived dopamine D2/D3 receptor agonist used in research. It shows high affinity for D2 receptors (pKi ≈ 8.05) and is provided as both a solid and solution format for laboratory and analytical applications.
Catalog No. 50-002-64443 Supplier Medchemexpress LLC Supplier No. HY12705A5MG
Bromocriptine mesylate is the methanesulfonate salt of bromocriptine, a semisynthetic ergot-derived dopamine D2/D3 receptor agonist used in research. It shows high affinity for D2 receptors (pKi ≈ 8.05) and is provided as both a solid and solution format for laboratory and analytical applications.