Learn More
Cayman Chemical Bromocriptine (mesylate), Purity: ≥98%, CAS: 22260-51-1, 100 mg
SDP
Catalog No. 502968519
Encompass_Preferred
A dopamine receptor agonist (Kis = 1,659, 12.2, 12.2, 59.7, and 1,691 nM for dopamine D1-5 receptors, respectively); also binds to 5-HT1A and 5-HT1D (Kis = 12.9 and 10.7 nM, respectively), as well as α1-adrenergic receptors (Kis = 1.12-4.17 nM); restores locomotor activity, without inducing dyskinesia, in a MPTP-induced macaque model of Parkinson’s disease at 5 mg/kg
Catalog No. 50-296-8519 Supplier Cayman Chemical Supplier No. 14598100 MG
A dopamine receptor agonist (Kis = 1,659, 12.2, 12.2, 59.7, and 1,691 nM for dopamine D1-5 receptors, respectively); also binds to 5-HT1A and 5-HT1D (Kis = 12.9 and 10.7 nM, respectively), as well as α1-adrenergic receptors (Kis = 1.12-4.17 nM); restores locomotor activity, without inducing dyskinesia, in a MPTP-induced macaque model of Parkinson’s disease at 5 mg/kg