Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bromocyclobutane 96.0+%, TCI America™
SDP

Catalog No. C07715G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C07715G 5 g
C07711G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C07715G Supplier TCI America Supplier No. C07715G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 4399-47-7
Molecular Formula C4H7Br
Molecular Weight (g/mol) 135.004
MDL Number MFCD00001317
InChI Key KXVUSQIDCZRUKF-UHFFFAOYSA-N
Synonym cyclobutyl bromide, cyclobutane, bromo, cyclobutylbromid, cyclobutylbromide, bromo cyclobutane, bromo-cyclobutane, bromocyclobutane, acmc-209jw1, ksc235o4p, inchi=1/c4h7br/c5-4-2-1-3-4/h4h,1-3h
PubChem CID 78110
IUPAC Name bromocyclobutane
SMILES C1CC(C1)Br

Specifications

Color Yellow
Boiling Point 108°C
Quantity 5 g
UN Number 1993
Formula Weight 135.00
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material Bromocyclobutane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.