Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Bromocyclohexane

Catalog No. 135194100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
13-519-4100G 100 g
13-519-41KG 1 kg
13-519-4500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 13-519-4100G Supplier Sigma Aldrich Supplier No. 135194100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Axial conformer of bromocyclohexane forms inclusion complexes with 9,9α-bianthryl and exhibits a weak 1,3 diaxial cl...h interaction
  • It participates in the alkylation reaction of of para-xylene in the presence of graphite as catalyst to yield hydrocarbon.bromocyclohexane has been used to study the ionization of equatorial and axial conformational isomers of the chair-bromocyclohexane using vacuum ultraviolet mass-analyzed threshold ionization spectroscopy

Specifications

CAS 108-85-0
Density 1.324 g/mL (at 25°C (literature))
Boiling Point 166°C to 167°C (lit.)
Molecular Formula C6H11Br
Linear Formula C6H11Br
Refractive Index n20/D 1.495 (literature)
MDL Number MFCD00003819
Quantity 100 g
Synonym Cyclohexyl bromide
Molecular Weight (g/mol) 163.06
Percent Purity 98%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.