Learn More
Medchemexpress LLC Bromocyclohexane-d11 | 35558-49-7 | MFCD00190385 | 98.1% | 174.12 | C6D11Br | 5 MG
SDP
Supplier:  Medchemexpress LLC HYW017209S5MG
Encompass_Preferred
Bromocyclohexane-d11 is a deuterium-labeled bromocyclohexane used as a tracer and internal standard in analytical chemistry. It has molecular formula C6D11Br, molecular weight 174.12, and is supplied as a colorless to light yellow liquid with reported purity ~98.1%. Typical applications include quantitative NMR, GC-MS, and LC-MS analyses.
  • Deuterium-labeled tracer for isotope dilution and tracing studies.
  • Suitable as an internal standard for quantitative NMR, GC-MS, and LC-MS.
  • High purity for reliable quantitative results.
  • Stable liquid form with defined density for accurate handling.
  • Available in small, preweighed quantities for analytical workflows.
Catalog No. 50-003-00572