Learn More
Medchemexpress LLC Bromocyclohexane-d11 | 35558-49-7 | MFCD00190385 | 98.1% | 174.12 g/mol | C6D11Br | 10 MG
SDP
Catalog No. 5000300574
Encompass_Preferred
Bromocyclohexane-d11 is a deuterium-labeled analogue of bromocyclohexane (C6D11Br, CAS 35558-49-7) used as a stable-isotope tracer and internal standard in analytical and mechanistic studies. It is supplied as a colorless to light yellow liquid with high deuterium incorporation for research applications.
  • Deuterium-labeled tracer for mass spectrometry and quantitative analyses.
  • High chemical purity and isotope enrichment for reliable standards.
  • Liquid form, colorless to light yellow, easy to handle and dispense.
  • Recommended storage at low temperature to maintain stability.
  • Available in small pack sizes suited to analytical workflows.
Catalog No. 50-003-00574 Supplier Medchemexpress LLC Supplier No. HYW017209S10MG
Bromocyclohexane-d11 is a deuterium-labeled analogue of bromocyclohexane (C6D11Br, CAS 35558-49-7) used as a stable-isotope tracer and internal standard in analytical and mechanistic studies. It is supplied as a colorless to light yellow liquid with high deuterium incorporation for research applications.
  • Deuterium-labeled tracer for mass spectrometry and quantitative analyses.
  • High chemical purity and isotope enrichment for reliable standards.
  • Liquid form, colorless to light yellow, easy to handle and dispense.
  • Recommended storage at low temperature to maintain stability.
  • Available in small pack sizes suited to analytical workflows.