Learn More
Medchemexpress LLC Bromocyclohexane-d11 | 35558-49-7 | MFCD00190385 | 98.1% | 174.12 g/mol | C6D11Br | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYW017209S
Encompass_Preferred
Bromocyclohexane-d11 is the deuterium-labeled isotopologue of bromocyclohexane, provided as a liquid research reagent. It is used as an internal standard and tracer in mass spectrometry, isotopic labeling, and mechanistic studies, offering reliable isotopic enrichment and chemical purity for quantitative analyses.
  • Deuterium-labeled reagent suitable for mass spectrometry internal standards.
  • High isotopic enrichment and chemical purity for accurate quantitation.
  • Liquid form simplifies handling and solution preparation.
  • Applicable to tracer and mechanistic studies in synthetic and analytical chemistry.
  • Characterized with molecular formula C6D11Br and molecular weight 174.12 g/mol.
  • Supplied with purity and documentation to support experimental use.
Catalog No. 50-003-03952