Learn More
Medchemexpress LLC Bromocyclohexane-d11 | 35558-49-7 | MFCD00190385 | 98.1% | 174.12 g/mol | C6D11Br | 50 MG
SDP
Supplier:  Medchemexpress LLC HYW017209S50MG
Encompass_Preferred
Bromocyclohexane-d11 is a deuterium-labeled analog of bromocyclohexane supplied for research use as a tracer and internal standard. It has formula C6D11Br and is suitable for quantitative analysis by NMR, GC-MS, and LC-MS. The material is provided with manufacturer-specified purity and storage recommendations for analytical workflows.
  • Deuterium-labeled for use as an internal standard in quantitative assays.
  • Compatible with NMR, GC-MS, and LC-MS analytical techniques.
  • Liquid form with defined molecular formula for accurate mass calculations.
  • High purity suitable for analytical applications.
  • Recommended storage conditions to preserve isotopic integrity.
Catalog No. 50-003-01863