Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bromodiphenylmethane, 90%

Catalog No. p-3598316
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC373461000 100 g
AC373465000 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC373461000 Supplier Thermo Scientific Chemicals Supplier No. 373461000
Only null left
Add to Cart

CAS: 776-74-9 | C13H11Br | 247.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 776-74-9
Molecular Formula C13H11Br
Molecular Weight (g/mol) 247.13
MDL Number MFCD00000134
InChI Key OQROAIRCEOBYJA-UHFFFAOYSA-N
Synonym bromodiphenylmethane, benzhydryl bromide, diphenylmethyl bromide, diphenylbromomethane, alpha-bromodiphenylmethane, bromo phenyl methyl benzene, benzene, 1,1'-bromomethylene bis, methane, bromodiphenyl, bromomethylene dibenzene, bromodiphenyl methane
PubChem CID 236603
IUPAC Name [bromo(phenyl)methyl]benzene
SMILES C1=CC=C(C=C1)C(C2=CC=CC=C2)Br

Specifications

Melting Point 33.0°C to 42.0°C
Boiling Point 184.0°C (20.0 mmHg)
Flash Point >110°C
Assay Percent Range 88% min. (GC)
Packaging Glass bottle
Linear Formula (C6H5)2CHBr
Quantity 100 g
Beilstein 05,592
Solubility Information Solubility in water: insoluble
Formula Weight 247.13
Percent Purity 90%
Chemical Name or Material Bromodiphenylmethane
Show More Show Less
Hazard Category GHS Signal Word: Danger
EINECSNumber 212-279-8

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.