Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bromodiphenylmethane 95%, Thermo Scientific™

Catalog No. AC106901000
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC106901000 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC106901000 Supplier Thermo Scientific Chemicals Supplier No. 106901000

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 776-74-9
Molecular Formula C13H11Br
Molecular Weight (g/mol) 247.135
InChI Key OQROAIRCEOBYJA-UHFFFAOYSA-N
Synonym bromodiphenylmethane, benzhydryl bromide, diphenylmethyl bromide, diphenylbromomethane, alpha-bromodiphenylmethane, bromo phenyl methyl benzene, benzene, 1,1'-bromomethylene bis, methane, bromodiphenyl, bromomethylene dibenzene, bromodiphenyl methane
PubChem CID 236603
IUPAC Name [bromo(phenyl)methyl]benzene
SMILES C1=CC=C(C=C1)C(C2=CC=CC=C2)Br

Specifications

Melting Point 36°C to 40°C
Color Brown-Orange to Orange-Red
Infrared Spectrum Authentic
Linear Formula (C6H5)2CHBr
Quantity 100 g
Beilstein 05, 592
Formula Weight 247.13
Percent Purity ≥93%
Physical Form Low Melting Solid
Chemical Name or Material Bromodiphenylmethane, 95%
Hazard Category GHS Signal Word: Danger
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.