Learn More
Apexbio Technology LLC Bromodomain Inhibitor, (+)-JQ1 1268524-70-4 10mM (in 1mL DMSO)
SDP
Catalog No. 5000063825
Encompass_Preferred
( )-JQ1 is a selective small-molecule inhibitor targeting BET (bromodomain and extraterminal) protein family members particularly BRD4 It competitively binds the acetyl-lysine recognition domain of BRD4 bromodomains 1 and 2 with reported Kd values of approximately 50 nM and 90 nM respectively By selectively inhibiting BET proteins ( )-JQ1 disrupts transcriptional regulatory processes essential in cancer biology rendering it a useful research tool for examining chromatin remodeling and gene expression patterns associated with tumor pathogenesis Additionally ( )-JQ1 specifically targets the testis-specific bromodomain protein BRDT enabling investigations into male reproductive biology by reversibly impairing spermatogenesis
Catalog No. 50-000-63825 Supplier Apexbio Technology LLC Supplier No. A19105.1
 Restricted Item
Add to Cart
Add to Cart
( )-JQ1 is a selective small-molecule inhibitor targeting BET (bromodomain and extraterminal) protein family members particularly BRD4 It competitively binds the acetyl-lysine recognition domain of BRD4 bromodomains 1 and 2 with reported Kd values of approximately 50 nM and 90 nM respectively By selectively inhibiting BET proteins ( )-JQ1 disrupts transcriptional regulatory processes essential in cancer biology rendering it a useful research tool for examining chromatin remodeling and gene expression patterns associated with tumor pathogenesis Additionally ( )-JQ1 specifically targets the testis-specific bromodomain protein BRDT enabling investigations into male reproductive biology by reversibly impairing spermatogenesis