Learn More
Medchemexpress LLC HY-128703 5mg Medchemexpress, Bromodomain inhibitor-8 CAS:1300031-70-2 Purity:>98%
SDP
Catalog No. 501932630
Encompass_Preferred
Medchemexpress, HY-128703 5mg Bromodomain inhibitor-8 CAS:1300031-70-2 Bromodomain inhibitor-8 (Intermediate 21) is a BET bromodomain inhibitor for treating autoimmune and inflammatory diseases [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2630 Supplier Medchemexpress LLC Supplier No. HY1287035MG
Medchemexpress, HY-128703 5mg Bromodomain inhibitor-8 CAS:1300031-70-2 Bromodomain inhibitor-8 (Intermediate 21) is a BET bromodomain inhibitor for treating autoimmune and inflammatory diseases [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.