Learn More
Cayman Chemical Bromo-DragonFLY (hydrochloride), Purity: ≥98%, CAS: 219986-78-4, 10 mg
SDP
Catalog No. 502952208
Encompass_Preferred
An analytical reference standard categorized as a phenethylamine; has been associated with fatalities and acute overdoses; induces the head-twitch response in mice, indicating hallucinogenic potential; intended for research and forensic applications
Catalog No. 50-295-2208 Supplier Cayman Chemical Supplier No. 1156110 MG
An analytical reference standard categorized as a phenethylamine; has been associated with fatalities and acute overdoses; induces the head-twitch response in mice, indicating hallucinogenic potential; intended for research and forensic applications