Learn More
Cayman Chemical Bromoenol lactone, Purity: ≥95%, CAS: 88070-98-8, 50 mg
SDP
Catalog No. 502896318
Encompass_Preferred
BEL is a selective, irreversible, mechanism-based inhibitor of iPLA2 with a Ki value of 180 nM.{1631} It inhibits macrophage iPLA2 in a concentration-dependent manner with an IC50 value of 60 nM.{1291,1770}
Catalog No. 50-289-6318 Supplier Cayman Chemical Supplier No. 7070050 MG
BEL is a selective, irreversible, mechanism-based inhibitor of iPLA2 with a Ki value of 180 nM.{1631} It inhibits macrophage iPLA2 in a concentration-dependent manner with an IC50 value of 60 nM.{1291,1770}