Learn More
Apexbio Technology LLC Bromoenol lactone 88070-98-8 25mg
SDP
Catalog No. 5000091547
Encompass_Preferred
Bromoenol lactone (CAS 88070-98-8) is an irreversible inhibitor of cytosolic calcium-independent phospholipase A2 (iPLA2) an enzyme involved in the regulation of arachidonic acid release and lysophospholipid production iPLA2 activity is associated with phospholipid remodeling and the generation of signaling molecules that modulate processes such as eicosanoid synthesis insulin secretion apoptosis and cellular proliferation In cellular assays bromoenol lactone induces apoptosis in various cell lines and promotes proteolysis of procaspase-9 and procaspase-3 It also inhibits phosphatidic acid phosphohydrolase activity and disrupts triacylglycerol biosynthesis in macrophage models Bromoenol lactone is widely utilized in studies investigating lipid signaling cell death mechanisms and metabolic pathways
Catalog No. 50-000-91547 Supplier Apexbio Technology LLC Supplier No. C508125
Bromoenol lactone (CAS 88070-98-8) is an irreversible inhibitor of cytosolic calcium-independent phospholipase A2 (iPLA2) an enzyme involved in the regulation of arachidonic acid release and lysophospholipid production iPLA2 activity is associated with phospholipid remodeling and the generation of signaling molecules that modulate processes such as eicosanoid synthesis insulin secretion apoptosis and cellular proliferation In cellular assays bromoenol lactone induces apoptosis in various cell lines and promotes proteolysis of procaspase-9 and procaspase-3 It also inhibits phosphatidic acid phosphohydrolase activity and disrupts triacylglycerol biosynthesis in macrophage models Bromoenol lactone is widely utilized in studies investigating lipid signaling cell death mechanisms and metabolic pathways