Learn More
Medchemexpress LLC Bromoenol lactone | 88070-98-8 | MFCD00270871 | ≥98.0% | 317.18 g/mol | C16H13BrO2 | 1 ML
SDP
Catalog No. 5000241403
Encompass_Preferred
Bromoenol lactone is an irreversible, suicide-type inhibitor of calcium-independent phospholipase A2 (iPLA2β). It is provided as a concentrated solution (commonly 10 mM in DMSO) for biochemical and cell-based research, with a reported IC50 of approximately 7 μM.
  • Irreversible inhibitor of calcium-independent phospholipase A2 (iPLA2β).
  • Reported IC50 of approximately 7 μM.
  • Supplied as a 10 mM solution in DMSO for laboratory use.
  • High purity: ≥98.0% (TLC).
  • Chemical formula C16H13BrO2, molecular weight 317.18 g/mol.
  • For research use only; not for human therapeutic use.
Catalog No. 50-002-41403 Supplier Medchemexpress LLC Supplier No. HY10741110MM 1ML
Bromoenol lactone is an irreversible, suicide-type inhibitor of calcium-independent phospholipase A2 (iPLA2β). It is provided as a concentrated solution (commonly 10 mM in DMSO) for biochemical and cell-based research, with a reported IC50 of approximately 7 μM.
  • Irreversible inhibitor of calcium-independent phospholipase A2 (iPLA2β).
  • Reported IC50 of approximately 7 μM.
  • Supplied as a 10 mM solution in DMSO for laboratory use.
  • High purity: ≥98.0% (TLC).
  • Chemical formula C16H13BrO2, molecular weight 317.18 g/mol.
  • For research use only; not for human therapeutic use.