Learn More
Medchemexpress LLC Bromoenol lactone | 88070-98-8 | MFCD00270871 | ≥98.0% | 317.18 g/mol | C16H13BrO2 | 10 MG
SDP
Catalog No. 5000241402
Encompass_Preferred
Bromoenol lactone is a suicide-based, irreversible inhibitor of calcium-independent phospholipase A2 (iPLA2β) used in biochemical research. It inhibits iPLA2β with an IC50 of approximately 7 μM and has been reported to block antigen-stimulated mast cell exocytosis without affecting Ca2+ influx. The compound is supplied as a solid for laboratory preparation of stock solutions.
  • Irreversible mechanism-based inhibitor of iPLA2β.
  • Reported IC50 of approximately 7 μM.
  • Useful for studying phospholipase A2-mediated signaling and mast cell exocytosis.
  • Available as a solid suitable for solution preparation.
  • Molecular weight 317.18 g/mol; molecular formula C16H13BrO2.
Catalog No. 50-002-41402 Supplier Medchemexpress LLC Supplier No. HY10741110MG
Bromoenol lactone is a suicide-based, irreversible inhibitor of calcium-independent phospholipase A2 (iPLA2β) used in biochemical research. It inhibits iPLA2β with an IC50 of approximately 7 μM and has been reported to block antigen-stimulated mast cell exocytosis without affecting Ca2+ influx. The compound is supplied as a solid for laboratory preparation of stock solutions.
  • Irreversible mechanism-based inhibitor of iPLA2β.
  • Reported IC50 of approximately 7 μM.
  • Useful for studying phospholipase A2-mediated signaling and mast cell exocytosis.
  • Available as a solid suitable for solution preparation.
  • Molecular weight 317.18 g/mol; molecular formula C16H13BrO2.