Learn More
Medchemexpress LLC Bromoenol lactone | 88070-98-8 | MFCD00270871 | ≥98.0% | 317.18 g/mol | C16H13BrO2 | 5 MG
SDP
Catalog No. 5000241286
Encompass_Preferred
Bromoenol lactone is a small-molecule, suicide-based irreversible inhibitor of calcium-independent phospholipase A2 (iPLA2). It is commonly used in biochemical and cellular research to probe iPLA2-related signaling, study lipid metabolism, and inhibit enzyme activity in assays and cell models.
  • Irreversible inhibition of calcium-independent phospholipase A2 (iPLA2).
  • Applicable to enzymatic assays and cell-based studies.
  • Used to investigate lipid metabolism and signaling pathways.
  • Provided in small research-scale quantities for laboratory use.
Catalog No. 50-002-41286 Supplier Medchemexpress LLC Supplier No. HY1074115MG
Bromoenol lactone is a small-molecule, suicide-based irreversible inhibitor of calcium-independent phospholipase A2 (iPLA2). It is commonly used in biochemical and cellular research to probe iPLA2-related signaling, study lipid metabolism, and inhibit enzyme activity in assays and cell models.
  • Irreversible inhibition of calcium-independent phospholipase A2 (iPLA2).
  • Applicable to enzymatic assays and cell-based studies.
  • Used to investigate lipid metabolism and signaling pathways.
  • Provided in small research-scale quantities for laboratory use.