Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Bromoenol lactone | 88070-98-8 | MFCD00270871 | 99.6% | 317.18 g/mol | C16H13BrO2 | 50 MG
SDP

Catalog No. 5000239940
Encompass_Preferred

Bromoenol lactone is a mechanism-based, irreversible inhibitor of calcium-independent phospholipase A2 (iPLA2). Used as a research reagent, it is employed to probe iPLA2-mediated lipid metabolism and signaling in enzymatic and cell-based studies. Supplied as a high-purity small molecule for laboratory use.

  • Mechanism-based irreversible inhibitor of calcium-independent phospholipase A2 (iPLA2).
  • High purity (approximately 99.6%).
  • Molecular weight 317.18 g/mol.
  • Suitable for enzymatic and cell-based assays.
  • Soluble in common organic solvents such as DMSO.
  • Available in milligram pack sizes for research use.

Catalog No. 50-002-39940 Supplier Medchemexpress LLC Supplier No. HY10741150MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Bromoenol lactone is a mechanism-based, irreversible inhibitor of calcium-independent phospholipase A2 (iPLA2). Used as a research reagent, it is employed to probe iPLA2-mediated lipid metabolism and signaling in enzymatic and cell-based studies. Supplied as a high-purity small molecule for laboratory use.

  • Mechanism-based irreversible inhibitor of calcium-independent phospholipase A2 (iPLA2).
  • High purity (approximately 99.6%).
  • Molecular weight 317.18 g/mol.
  • Suitable for enzymatic and cell-based assays.
  • Soluble in common organic solvents such as DMSO.
  • Available in milligram pack sizes for research use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.